Learn More
R&D Systems™ Human CD4+/CD45RO- Naive T Cell Enrichment Column, 4 Pack
Description
The resulting column eluate is a highly enriched T cell subset population with minimal contamination of either CD45RO+ or CD8+ cells
Recovery of CD4+/45RO- cells from all available CD4+/45RO- cells loaded ranged between 13% and 26%
Purity (CD4+/45RO-) of recovered cells ranged between 93% and 96% with <1% CD4+/45RO+ cells detectable among all cells recovered
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Quantity | 4/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.