Learn More
R&D Systems™ Human CD4+ T Cell Enrichment Column, Mini
Description
CD4 is an approximately 55kDa type I transmembrane glycoprotein that is expressed predominantly on thymocytes and a subset of mature T cells. In humans, CD4 is also expressed to a lesser extent on monocytes and macrophage related cells.
The resulting column eluate is a highly enriched CD4+ cell subset population with minimal depletion of the selected T cells
Minimal numbers of B cells, monocytes, or non-selected T cell subsets
Recovery of CD3+/4+ cells from all available CD3+/4+ cells ranged between 32% and 53%
Purity of recovered cell ranging between 88% and 95% with no detectable CD3+/8+ cells among recovered cells
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Quantity | 4/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.