Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CD40 Ligand/TNFSF5 (aa 108-261) Protein
SDP

Catalog No. 6245CL050CF Shop All Novus Biologicals Products
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in NaH2PO4, NaCl and EDTA with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in NaH2PO4, NaCl and EDTA.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
6245CL050CF Lyophilized from a 0.2 μm filtered solution in NaH2PO4, NaCl and EDTA.
6245CL050 Lyophilized from a 0.2 μm filtered solution in NaH2PO4, NaCl and EDTA with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 6245CL050CF Supplier R&D Systems™ Supplier No. 6245CL050/CF

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A New rh CD40 Ligand is Available! ∼1000 fold better activity; HEK293 expressed; HA tag!

Measured in a cell proliferation assay using B cell-enriched peripheral blood mononuclear cells (PBMC) in the presence of IL-4. Spriggs, M.K. et al. (1992) J. Exp. Med. 176:1543. The ED50 for this effect is 1-3.00 μg/mL in the presence of 20 ng/mL of recombinant human IL-4.

Specifications

Accession Number P29965
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in NaH2PO4, NaCl and EDTA.
Gene ID (Entrez) 959
Molecular Weight (g/mol) M.W. (Observed): 18 kDa, reducing conditions; M.W. (theoretical): 16.9 kDa (monomer)
Name CD40 Ligand/TNFSF5 aa 108-261
Quantity 50 μg
Source E. coli-derived human CD40 Ligand/TNFSF5 protein Glu108-Leu261, with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 3 months, 2°C to 8°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol CD40LG
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in PBS.
Product Line R&D Systems
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein CD40 Ligand/TNFSF5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.