Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CD40 Ligand/TNFSF5 (Histidine-tagged)

Catalog No. 2706CL025CF
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in HEPES, NaCl and EDTA with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in HEPES, NaCl and EDTA.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
2706CL025CF Lyophilized from a 0.2 μm filtered solution in HEPES, NaCl and EDTA.
2706CL025 Lyophilized from a 0.2 μm filtered solution in HEPES, NaCl and EDTA with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 2706CL025CF Supplier R&D Systems™ Supplier No. 2706CL025/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

A New rh CD40 Ligand is Now Available! ∼10 fold better activity; HEK293 expressed; HA tag!

Measured in a cell proliferation assay using enriched human B cells in the presence of IL-4. Spriggs, M.K. et al. (1992) J. Exp. Med. 176:1543. The ED50 for this effect is 2-12 ng/mL in the presence of 10 μg/mL of a cross-linking antibody, Mouse Anti-polyHistidine Monoclonal Antibody (Catalog # MAB050 ) and 20 ng/mL of recombinant human IL-4.

Specifications

Accession Number P29965
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in HEPES, NaCl and EDTA.
Gene ID (Entrez) 959
Molecular Weight (g/mol) M.W. (Observed): 18 kDa, reducing conditions; M.W. (theoretical): 17.7 kDa (monomer)
Name CD40 Ligand/TNFSF5 (Histidine-tagged)
Quantity 25 μg
Source E. coli-derived human CD40 Ligand/TNFSF5 protein Glu108-Leu261, with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution.
Structural Form Homotrimer
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol CD40LG
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein CD40 Ligand/TNFSF5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.