Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CD40 Ligand/TNFSF5 (HEK293-expressed)

Catalog No. 6420CL025CF
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS and EDTA with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS and EDTA.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
6420CL025CF Lyophilized from a 0.2 μm filtered solution in PBS and EDTA.
6420CL025 Lyophilized from a 0.2 μm filtered solution in PBS and EDTA with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 6420CL025CF Supplier R&D Systems™ Supplier No. 6420CL025/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using enriched human B cells in the presence of IL-4. Spriggs, M.K. et al. (1992) J. Exp. Med. 176:1543. The ED50 for this effect is 0.2-1.2 ng/mL in the presence of 20 ng/mL of Recombinant Human IL-4 (Catalog # 204-IL ) and a cross-linking antibody, Mouse Anti-Hemagglutinin/HA Peptide Monoclonal Antibody (Catalog # MAB060 ).

Specifications

Accession Number P29965
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and EDTA.
Gene ID (Entrez) 959
Molecular Weight (g/mol) M.W. (Observed): 22-26 kDa, reducing condtions; M.W. (theoretical): 21.6 kDa
Name CD40 Ligand/TNFSF5
Quantity 25 μg
Source Human embryonic kidney cell, HEK293-derived human CD40 Ligand/TNFSF5 protein Hemagglutinin tag YPYDVPDYA GCN4-IZ (Met1-Ile29) GGGSGGGSGGGS Human CD40 Ligand (Met113-Leu261) (N-terminus) Accession # P29965 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol CD40LG
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein CD40 Ligand/TNFSF5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.