Learn More
R&D Systems™ Human CD8+/45RA- Memory T Cell Enrichment Column
Description
The resulting column eluate is a highly enriched T cell subset population with minimal contamination of either CD45RO+ or CD4+ cells
Virtually no B cells, monocytes, or non-selected T cell subsets
Recovery of CD8+/CD45RA- cells from all available CD8+/CD45RA- cells loaded ranged between 4% and 14%
Purity (CD8+/CD45RA-) of recovered cells ranged between 80% and 90%, with no detectable CD8+/CD45RA+ or CD3+/CD4+ cells among recovered cells
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Quantity | 4/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.