Learn More
R&D Systems™ Human CD8+ T Cell Enrichment Column, Small
Description
CD8 is a heterodimeric glycoprotein consisting of an alpha and beta chain. It is expressed on cytolytic T cells and functions in conjunction with the T cell receptor in the recognition of MHC/peptide complexes.
The resulting column eluate is a highly enriched T cell subset population with minimal depletion of the selected T cells
Minimal B cells, monocytes, or non-selected T cell subsets
Recovery of CD3+/CD8+ cells from all available CD3+/CD8+ cells loaded ranged between 29% and 37%
Purity of recovered cells ranged between 83% and 88% with no detectable CD3+/CD4+ in the recovered cells
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Quantity | 4/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.