Learn More
R&D Systems™ Human Chemokine Sampler Packs T/DC Differentiation Protein Sampler Pack
Catalog No. SMPK6
Change view
Click to view available options
1 Pack
Catalog No.
SMPK6 1 Pack
1 options
This item is not returnable. View return policy
Catalog No. SMPK6 Supplier R&D Systems™ Supplier No. SMPK6
This item is not returnable. View return policy
Regulatory Status: RUO
Specifications
Name Chemokine Sampler Packs T/DC Differentiation
Quantity 1 Pack
Storage Requirements Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Conjugate Unconjugated