Learn More
R&D Systems™ Human Choline Kinase beta Recombinant Protein
Description
Source: E. coli-derived, Arg35-Ser395, with an N-terminal Met and a C-terminal 6-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 Âμg per lane.
Molecular Weight: 43 kDa
Formulation: Supplied as a 0.2μm filtered solution in Tris, NaCl and DTT.
Entrez Gene ID: 1120
Regulatory Status: RUO
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris, NaCl, TCEP and Glycerol. |
| Gene ID (Entrez) | 1120 |
| Molecular Weight (g/mol) | 43 kDa |
| Name | Choline Kinase beta |
| Quantity | 20 μg |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.