Learn More
R&D Systems™ Human Coagulation Factor II/Thrombin Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in MES, NaCl and Brij-35. |
| Gene ID (Entrez) | 2147 |
| Name | Coagulation Factor II/Thrombin |
| Quantity | 200 μg |
| Source | Human plasma-derived Coagulation Factor II/Thrombin protein The human plasma used for the isolation of this product was certified by the supplier to be non-reactive for HIV-1/2 and HBsAg negative at the donor level. Human blood products should always be treated in accordance with universal handling precautions. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | pro form |
| Conjugate | Unconjugated |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.