Learn More
R&D Systems™ Human Common beta Chain Fc Chimera Recombinant Protein
Description
Source: Mouse myeloma cell line, NS0-derived, GlMet1-Trp443 with a C-terminal Fc
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 74.6 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 1439
Regulatory Status: RUO
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 1439 |
| Molecular Weight (g/mol) | 74.6 kDa (monomer) |
| Name | Common beta Chain Fc Chimera |
| Quantity | 50 μg |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.