Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CTLA-4 Fc Chimera His-tag Protein
SDP

Catalog No. 325CT200 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
325CT200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 325CT200 Supplier R&D Systems™ Supplier No. 325CT200
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

New rh CTLA-4 Available! Increased activity; Enhanced purity; Lower endotoxin specification; CHO expressed!

Measured by its ability to inhibit IL-2 secretion by stimulated Jurkat human acute T cell leukemia cells. Linsley, P.S. et al. (1991) J. Exp. Med. 174:561. The ED50 for this effect is 0.1-0.4 μg/mL when stimulated with 1 μg/mL Recombinant Human B7-1/CD80 Fc Chimera (Catalog # 140-B1 ) in the presence of PHA.

Specifications

Accession Number Q6GR94
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 1493
Molecular Weight (g/mol) M.W. (theoretical): 40 kDa (monomer)
Name CTLA-4 Fc Chimera
Quantity 200 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human CTLA-4 protein Human CTLA-4 (Ala37-Phe162) (N-terminus) Accession # Q6GR94 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag(C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol CTLA4
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in sterile PBS containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein CTLA-4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.