Learn More
R&D Systems™ Human Di-Ubiquitin/Ub2 Non-hydrolyzable (K63), Agarose Recombinant Protein
Description
Specifications
Specifications
| Accession Number | P0CG47 |
| Formulation | No Formulation Info in Bulk Inventory |
| Gene ID (Entrez) | 7314 |
| Name | Di-Ubiquitin/Ub2 Non-hydrolyzable (K63), Agarose |
| Quantity | 250 μL |
| Source | E. coli-derived human Di-Ubiquitin protein Each Ubiquitin contains a Pro substitution at position 73 |
| Storage Requirements | Do not freeze. 3 months from date of receipt, 2 to 8° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. |
| Conjugate | Agarose |
| Recombinant | Recombinant |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.