Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human EGF DuoSet ELISA
SDP

Catalog No. DY23605 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit (for 5 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY23605 1 Kit (for 5 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY23605 Supplier R&D Systems™ Supplier No. DY23605

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Human EGF Capture Antibody; Human EGF Detection Antibody; Human EGF Standard; Streptavidin-HRP

Human EGF DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human EGF DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human EGF.

Specifications

Assay Range 3.91 to 250 pg/mL
Conjugate Biotin
Product Type Solid Phase Sandwich ELISA
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 1950
Gene Symbol EGF
Quantity 1 Kit (for 5 Plates)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), hEGF, HOMG4, pro-epidermal growth factor, URG, Urogastrone
Target EGF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.