Learn More
R&D Systems™ Human Erythrocyte Lysing Kit

Description
- Gently lyses erythrocytes
- Stabilizes leukocytes for analysis by flow cytometry
- Maintains natural light scattering and fluorescent staining properties of leukocytes
- Maintains viability of leukocytes for subsequent tissue culture manipulation
Specifications
Specifications
| Includes | H-Lyse Buffer Concentrate (10X); Wash Buffer Concentrate (10X); Fixative Concentrate (10X) |
| Product Type | Cell Lysis Kit |
| Content And Storage | Store at 20°C to 25°C |
| For Use With (Application) | Removal of erythrocytes (red blood cells) from whole blood |
| Sufficient For | 250 mL of whole blood |
| Quantity | 1 Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.