Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Erythropoietin/EPO (Tissue Culture Grade)
Shop All Novus Biologicals Products

Description
The Recombinant Human Erythropoietin (Tissue Culture Grade) is derived from CHO. The Recombinant Human Erythropoietin (Tissue Culture Grade) has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | NP_000790.2 |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 2056 |
| Molecular Weight (g/mol) | 21 kDa |
| Name | Erythropoietin/EPO |
| Quantity | 500 U |
| Source | Chinese Hamster Ovary cell line, CHO-derived human Erythropoietin/EPO protein Ala28-Arg193 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2 |
| Conjugate | Unconjugated |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?