Learn More
R&D Systems™ Human FAT10 Agarose Recombinant Protein
Description
Specifications
Specifications
| Formulation | Formulation cannot be determined without the BulkLotPrefix |
| Gene ID (Entrez) | 10537 |
| Name | FAT10 Agarose |
| Quantity | 500μL |
| Source | E. coli-derived human FAT10 protein |
| Storage Requirements | Do not freeze. 3 months from date of receipt, 2 to 8° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. |
| Conjugate | Agarose |
| Recombinant | Recombinant |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.