Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human FGF-4 (aa 71-206) Protein

Catalog No. 7460F4025
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 and EDTA.
Lyophilized from a 0.2 μm filtered solution in MOPS, Na2SO4 and EDTA with BSA as a carrier protein.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
7460F4025 Lyophilized from a 0.2 μm filtered solution in MOPS, Na2SO4 and EDTA with BSA as a carrier protein.
7460F4025CF Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 and EDTA.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 7460F4025 Supplier R&D Systems™ Supplier No. 7460F4025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749. The ED50 for this effect is 0.25-1.25 ng/mL.

Specifications

Accession Number P08620
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in MOPS, Na2SO4 and EDTA with BSA as a carrier protein.
Gene ID (Entrez) 2249
Molecular Weight (g/mol) M.W. (Observed): 14 kDa, reducing conditions; M.W. (theoretical): 15.2 kDa
Name FGF-4 (aa 71-206)
Quantity 25 μg
Source E. coli-derived human FGF-4 protein Ser71-Leu206 with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Monomer
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol FGF4
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein FGF-4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.