Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human FGF-4 (aa 71-206) Protein
SDP

Catalog No. 7460F4025CF Shop All Novus Biologicals Products
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 and EDTA.
Lyophilized from a 0.2 μm filtered solution in MOPS, Na2SO4 and EDTA with BSA as a carrier protein.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
7460F4025CF Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 and EDTA.
7460F4025 Lyophilized from a 0.2 μm filtered solution in MOPS, Na2SO4 and EDTA with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 7460F4025CF Supplier R&D Systems™ Supplier No. 7460F4025/CF

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human FGF-4 (aa 71-206) Protein is derived from E. coli. The Recombinant Human FGF-4 (aa 71-206) Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 and EDTA.
Gene ID (Entrez) 2249
Molecular Weight (g/mol) 15.2 kDa
Name FGF-4 (aa 71-206)
Quantity 25 μg
Source E. coli-derived human FGF-4 protein Ser71-Leu206 with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Monomer
Conjugate Unconjugated
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.