Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
The Human Fibronectin Protein is derived from Human Plasma. The Human Fibronectin Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris-HCl, NaCl and Urea. |
| Gene ID (Entrez) | 2335 |
| Name | Fibronectin |
| Quantity | 2 mg |
| Source | Human plasma-derived Fibronectin protein The human plasma used for the isolation of this product was certified by the supplier to be HIV-1 and HBsAg negative at the time of shipment. Human blood products should always be treated in accordance with universal handling precautions. |
| Storage Requirements | 6 months from date of receipt, 2 to 8° C as supplied. 3 months, 2 to 8° C under sterile conditions after opening. Avoid vortexing and excessive agitation. |
| Conjugate | Unconjugated |
| Purity or Quality Grade | 90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
