Learn More
R&D Systems™ Human Galectin-2 Recombinant Protein
Description
Source: E. coli-derived, Met1-Glu132
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and Betamercaptoethanol.
Entrez Gene ID: 3957
Regulatory Status: RUO
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS and Betamercaptoethanol with BSA as a carrier protein. |
| Gene ID (Entrez) | 3957 |
| Molecular Weight (g/mol) | 14 kDa |
| Name | Galectin-2 |
| Quantity | 50 μg |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.