Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human GST-AMSH/STAMBP Protein, CF

Catalog No. E549050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E549050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E549050 Supplier R&D Systems™ Supplier No. E549050
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Recombinant Human GST-AMSH is a Ubiquitin-specific deconjugating enzyme. Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human GST-AMSH concentration of 100-500 nM. GST-AMSH activity is partially or fully inhibited by DTT at concentrations above 1 mM.

Specifications

Accession Number O95630
For Use With (Application) Bioactivity
Formulation X mg/mL (X μM) in 50 mM HEPES pH 8.0, 10% Glycerol, 150 mM NaCl, 1 mM TCEP
Gene ID (Entrez) 10617
Molecular Weight (g/mol) M.W. (theoretical): 75 kDa
Name GST AMSH/STAMBP
Quantity 50 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human AMSH/STAMBP protein Contains an N-terminal GST (glutathione S-transferase) tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Symbol STAMBP
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Protein AMSH/STAMBP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.