Learn More
R&D Systems™ Human GST NEDD8 Activating Enzyme (APPBP1/UBA3) Recombinant Protein
Description
Specifications
Specifications
| Accession Number | Q13564, Q8TBC4 |
| Formulation | Formulation cannot be determined without the BulkLotPrefix |
| Gene ID (Entrez) | 8883 |
| Molecular Weight (g/mol) | Theoretical Mole. Wt.: 86 kDa (GST-APPBP1) 48 kDa (UBA3) |
| Name | GST NEDD8 Activating Enzyme (APPBP1/UBA3) |
| Quantity | 50 μg |
| Source | E. coli-derived human NEDD8 Activating Enzyme (APPBP1/UBA3) protein Contains an N-terminal GST (glutathione S-transferase) tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.