Learn More
R&D Systems™ Recombinant Human GST-UbcH2/UBE2H Protein, CF
Description
Specifications
Specifications
| Accession Number | P62256 |
| For Use With (Application) | Enzyme Activity |
| Formulation | X mg/mL (XμM) in 50 mM HEPES, pH 8.0, 100 mM NaCl, and 10% Glycerol (v/v) |
| Gene ID (Entrez) | 7328 |
| Molecular Weight (g/mol) | M.W. (theoretical): 46 kDa |
| Name | GST UbcH2/UBE2H |
| Quantity | 100 μg |
| Source | E. coli-derived human UbcH2/UBE2H protein Contains an N-terminal GST (glutathione S-transferase) tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Gene Symbol | UBE2H |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.