Learn More
R&D Systems™ Human HA GATE-16 Vinyl Sulfone C-Terminal Derivative Recombinant Protein
Description
Specifications
Specifications
| Formulation | Formulation cannot be determined without the BulkLotPrefix |
| Gene ID (Entrez) | 11345 |
| Name | HA GATE-16 Vinyl Sulfone C-Terminal Derivative |
| Quantity | 25μg |
| Source | E. coli-derived human GATE-16 protein |
| Storage Requirements | Store the unopened product at -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Conjugate | Vinyl Sulfone |
| Recombinant | Recombinant |
| Purity or Quality Grade | 90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.