Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human His6-ATG7 Isoform 1 Protein, CF

Catalog No. E318050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E318050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E318050 Supplier R&D Systems™ Supplier No. E318050

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Recombinant Human His6-ATG7 (Apg7) is a member of the Apg-activating (E1) enzyme family that is required for the first step of the enzymatic cascade that subsequently utilizes a Apg-conjugating (E2) enzyme and a Apg ligase (E3) to conjugate Apg to substrate proteins. Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human His6-ATG7 (Apg7) concentration of 50-200 nM.

Specifications

Accession Number O95352
For Use With (Application) Bioactivity
Formulation X mg/mL (X μM) in 50 mM HEPES pH 7.0, 150 mM NaCl, 2 mM TCEP, 10% glycerol
Gene ID (Entrez) 10533
Molecular Weight (g/mol) M.W. (theoretical): 80 kDa
Name His6 ATG7 Isoform 1
Quantity 50 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human ATG7 protein Contains an N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Symbol ATG7
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Protein ATG7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.