Learn More
R&D Systems™ Human His6 GATE-16 Rhodamine Red Recombinant Protein
Description
Specifications
Specifications
| Formulation | X mg/mL (X μM) in 50 mM Hepes pH 7.5, 100 mM NaCl.Actual concentration varies with lot number. |
| Gene ID (Entrez) | 11345 |
| Name | His6 GATE-16 Rhodamine Red |
| Quantity | 50μg |
| Source | E. coli-derived human GATE-16 protein Contains an N-terminal 6-His tag |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Conjugate | Rhodamine |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by PAGE. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.