Learn More
R&D Systems™ Human His6 PA28 Activator gamma Subunit Recombinant Protein
Description
Specifications
Specifications
| Formulation | X mg/mL (X μM) in 50 mM HEPES pH 8.0, 500 mM NaCl, 0.1 mM EDTA, 1 mM TCEP |
| Gene ID (Entrez) | 10197 |
| Molecular Weight (g/mol) | 31 kDa (monomer);188 kDa (hexamer) |
| Name | His6 PA28 Activator gamma Subunit |
| Quantity | 100 μg |
| Source | E. coli-derived human PA28 Activator gamma Subunit protein Contains an N-terminal MSYY and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.