Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human His6 UBE2J1 Recombinant Protein

Catalog No. E2750100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E2750100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E2750100 Supplier R&D Systems™ Supplier No. E2750100
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human His6-UBE2J1 Protein is derived from E. coli. The Recombinant Human His6-UBE2J1 Protein has been validated for the following applications: Enzyme Activity.

Specifications

Accession Number Q9Y385
Formulation X mg/mL (X μM) in 50 mM HEPES pH 8.0, 200 mM NaCl, 10% Glycerol (v/v), 1 mM TCEP
Gene ID (Entrez) 51465
Name His6 UBE2J1
Quantity 100 μg
Source E. coli-derived human UBE2J1 protein Contains an N-terminal 6-His tag and the C-terminal transmembrane and lumenal amino acid sequences have been removed
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Conjugate Unconjugated
Recombinant Recombinant
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.