Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human His6-UBE2Q2 Isoform 1 Protein, CF

Catalog No. E2745100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E2745100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E2745100 Supplier R&D Systems™ Supplier No. E2745100
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Recombinant Human His6-UBE2Q2-1 is a member of the Ubiquitin-conjugating (E2) enzyme family that receives Ubiquitin from a Ubiquitin-activating (E1) enzyme and subsequently interacts with a Ubiquitin ligase (E3) to conjugate Ubiquitin to substrate proteins. Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human His6-UBE2Q2-1 concentration of 0.1-1 μM.

Specifications

Accession Number Q8WVN8
For Use With (Application) Enzyme Activity
Formulation X mg/mL (X μM) in 50 mM HEPES, pH 7.5, 200 mM NaCl, 10% Glycerol (v/v), 2 mM DTT
Gene ID (Entrez) 92912
Molecular Weight (g/mol) M.W. (Observed): 44 kDa; M.W. (theoretical): 44 kDa
Name His6 UBE2Q2 Isoform 1
Quantity 100 μg
Source E. coli-derived human UBE2Q2 protein Contains an N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Gene Symbol UBE2Q2
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Protein UBE2Q2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.