Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human His6 Ubiquitin Mutant with K27 only Recombinant Protein

Catalog No. UMHK27001M
Change view
Click to view available options
1mg; Carrier Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
UMHK27001M 1mg; Carrier Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. UMHK27001M Supplier R&D Systems™ Supplier No. UMHK27001M

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human His6-Ubiquitin Mutant with K27 only is derived from E. coli. The Recombinant Human His6-Ubiquitin Mutant with K27 only has been validated for the following applications: Enzyme Activity.

Specifications

Accession Number P0CG47
Formulation No Formulation Info in Bulk Inventory
Gene ID (Entrez) 7314
Molecular Weight (g/mol) 9.8 kDa
Name His6 Ubiquitin Mutant with K27 only
Quantity 1mg
Source E. coli-derived human Ubiquitin protein Contains a 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute in deionized water or aqueous buffer.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.