Learn More
R&D Systems™ Human His6 Ubiquitin Mutant K27R Recombinant Protein
Description
Specifications
Specifications
| Accession Number | P0CG47 |
| Formulation | Formulation cannot be determined without the BulkLotPrefix |
| Gene ID (Entrez) | 7314 |
| Name | His6 Ubiquitin Mutant K27R |
| Quantity | 1mg |
| Source | E. coli-derived human Ubiquitin protein Contains a 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Reconstitution | Reconstitute in deionized water or aqueous buffer. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.