Learn More
R&D Systems™ Human His6 Ubiquitin Mutant with K63 only, N-Terminal Biotin Recombinant Protein
Description
Source: E. coli-derived, Contains a 6-His tag and a single N-terminal biotin
Purity: 95%, by PAGE.
Formulation: Formulation cannot be determined without the BulkLotPrefix
Entrez Gene ID: 7314
Regulatory Status: RUO
Specifications
Specifications
| Formulation | No Formulation Info in Bulk Inventory |
| Gene ID (Entrez) | 7314 |
| Name | His6 Ubiquitin Mutant with K63 only, N-Terminal Biotin |
| Quantity | 50μg |
| Conjugate | Biotin |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by PAGE. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.