Learn More
Description
Includes
- Microplate
- Biotinylated Detection Antibody
- Streptavidin conjugated to Alkaline Phosphatase
- Dilution Buffers
- Wash Buffer Concentrate
- BCIP/NBT Chromogen
- Positive Control
Complete ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. Kits are available for detection and enumeration of a single analyte or two analytes simultaneously.
- Detect and quantitate individual cells secreting
- High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
- No in vitro expansion of cells required
- High-throughput - ELISpot assays use only a small number of primary cells
Specifications
Specifications
| Includes | One 96-well PVDF-backed microplate coated w/polyclonal Ab for human IL-13, Biotinylated detection Ab, Streptavidin-AP, Buffers, BCIP/NBT, Positive control |
| Quantity | 1 x 96-well Plate |
| Sample Type | Whole Cells |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| For Use With (Application) | Diagnostics |
| Product Type | Human IL-13 ELISpot Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
