Learn More
Description
Complete ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. Kits are available for detection and enumeration of a single analyte or two analytes simultaneously.
- An economical alternative to ELISpot Kits
- Optimized capture and detection antibody pairings and recommended concentrations save lengthy development time
- Generic development protocols provide direction to start an optimization protocol
- Customize the assay to your specific needs
Module content:
- Capture Antibody
- Biotinylated-conjugated Detection Antibody
Specifications
Specifications
| Includes | Six 96-well PVDF-backed plates coated w/polyclonal Ab for human IL-2, Biotinylated detection Ab, Streptavidin-AP, Buffers, BCIP/NBT, Positive control |
| Quantity | 6 x 96-well Plates |
| Sample Type | Whole Cells |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Product Line | R&D Systems |
| For Use With (Application) | Diagnostics |
| Product Type | Human IL-2 ELISpot Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
