Learn More
Description
- The Human IL-5 ELISpot kit is a highly sensitive, microplate-based assay designed for the detection and enumeration of individual human IL-5 secreting cells.
- The assay employs a quantitative sandwich enzyme-linked immunosorbent assay (ELISA) technique.
- Assay Principle: Quantitative sandwich ELISA
- Sample Type: Whole cells
- Microplate: 96-well PVDF-backed microplate pre-coated with a monoclonal antibody specific for human IL-5
- Detection: Biotinylated detection antibody with Streptavidin conjugated to Alkaline Phosphatase (AP)
- Assay Length: 3 hour 15 min to 4 hour 30 min
- Key Contents: Human IL-5 microplate, 120xDetection Antibody Concentrate, 120xStreptavidin-AP Concentrate A, and lyophilized recombinant human IL-5 positive control
Specifications
Specifications
| Conjugate | AP |
| Cross Reactivity | Human |
| Quantity | 1 Kit |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Product Line | R&D Systems |
| Product Type | ELISpot Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
