Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-6 ELISpot Kit
SDP

Catalog No. EL206 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
6 x 96-well Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EL206 1 Kit
XEL206 6 x 96-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. EL206 Supplier R&D Systems™ Supplier No. EL206

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

highly sensitive, microplate-based assays for the detection of cytokine secreting cells

Product Features:
  • Detect and quantitate individual cells secreting
  • High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
  • No in vitro expansion of cells required
  • Hig

Specifications

Antigen IL-6
Conjugate AP
Cross Reactivity Human
Includes One 96-well PVDF-backed plate coated w/polyclonal Ab for human IL-6, Biotinylated detection Ab, Streptavidin-AP, Buffers, BCIP/NBT, Positive control
Quantity 1 Kit
Sample Type Whole Cells
Synonym B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Gene ID (Entrez) 3569
Gene Symbol IL6
Form Liquid
Product Line R&D Systems
For Use With (Application) Diagnostics
Host Species Human
Product Type Human IL-6 ELISpot Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.