Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-6/IL-6R alpha Complex DuoSet ELISA
SDP

Catalog No. DY813905 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY813905 5 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY813905 Supplier R&D Systems™ Supplier No. DY813905
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Capture Antibody, Detection Antibody, Recombinant Standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)

Human IL-6/IL-6R alpha Complex DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-6/IL-6R alpha Complex DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-6/IL-6R alpha Complex.

Specifications

Assay Sensitivity 0pg/mL
Conjugate Biotin
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Target Species Human
For Use With (Application) ELISA
Kit Contents Capture antibody, Detector antibody, Recombinant standard, Streptavidin-HRP
Quantity 5 Plate
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Target IL-6/IL-6R alpha Complex
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.