Learn More
R&D Systems™ Recombinant Human IL-7R alpha/CD127 Fc Chimera Protein, CF
Description
Specifications
Specifications
| Accession Number | BAG62793.1 |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 3575 |
| Molecular Weight (g/mol) | 54.5 kDa (monomer) |
| Name | IL-7R alpha/CD127 |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human IL-7 R alpha/CD127 protein, (N-terminus) Human IL-7 R alpha (Glu21-Lys261) Accession &Num; BAG62793.1 IIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Gene Alias | CD127, CD127 antigen, CD127ILRA, CDW127, IL-7 R alpha, IL-7 receptor subunit alpha, IL7R, IL7R alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.