Learn More
R&D Systems™ Human Integrin alpha V beta 1 Recombinant Protein
Description
Source: Chinese Hamster Ovary cell line, CHO-derived, Phe31-Val992 (aV subunit) & Gln21-Asp728 (beta1 subunit)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 115.4 kDa ( alphaV subunit) & 86.4 kDa (beta1 subunit)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3685
Regulatory Status: RUO
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 3685 |
| Molecular Weight (g/mol) | 115.4 kDa ( alphaV subunit) & 86.4 kDa (beta1 subunit) |
| Name | Integrin alpha V beta 1 |
| Quantity | 25 μg |
| Structural Form | Noncovalently-linked heterodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.