Learn More
R&D Systems™ Human ISG15/UCRP Agarose Recombinant Protein
Description
Specifications
Specifications
| Formulation | Supplied as a solution in 50mM HEPES pH 7.5, 250mM NaCl, 0.09% (w/v) Sodium Azide |
| Gene ID (Entrez) | 9636 |
| Name | ISG15/UCRP Agarose |
| Quantity | 500μL |
| Source | E. coli-derived human ISG15/UCRP protein |
| Storage Requirements | Do not freeze. 3 months from date of receipt, 2 to 8° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. |
| Conjugate | Agarose |
| Recombinant | Recombinant |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.