Learn More
R&D Systems™ Human ISG15/UCRP Fluorescein Recombinant Protein
Description
Specifications
Specifications
| Formulation | X mg/mL (X μM) in 50 mM HEPES, 100 mM NaCl, 1 mM DTT and 10% (v/v) glycerol, pH 7.5 |
| Gene ID (Entrez) | 9636 |
| Molecular Weight (g/mol) | 18 kDa (unlabeled), extent of labeling varies by lot |
| Name | ISG15/UCRP Fluorescein |
| Quantity | 50μg |
| Source | E. coli-derived human ISG15/UCRP protein |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Conjugate | Fluorescein |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.