Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human LDLR Quantikine ELISA Kit

Catalog No. DLDLR0
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DLDLR0 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DLDLR0 Supplier R&D Systems™ Supplier No. DLDLR0
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Cell Culture Supernates (50μL), Cell Lysates (50μL), Serum (10μL), Heparin Plasma (10μL), Urine (50μL)

Human LDLR Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human LDLR Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human LDLR in cell culture supernates (50 ul), cell lysates (50 ul), serum (10 ul), heparin plasma (10 ul), urine (50 ul).

Specifications

Assay Range 62.50 - 4000pg/mL
Assay Sensitivity 10.3pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Sample Type Cell Culture Supernatant, Cell Lysates, Plasma (Heparin), Serum, Urine
Specificity Natural and Recombinant Human LDL R. This assay also recognizes Recombinant Human LDL R/PCSK9 complex.
Target Species Human
For Use With (Application) ELISA
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Cell Lysates (50uL),Serum (10uL),Heparin Plasma (10uL),Urine (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym FH, FHC, LDL R, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3
Target LDLR
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.