Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human LSECtin/CLEC4G Protein, CF
SDP

Catalog No. 2947CL050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2947CL050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 2947CL050 Supplier R&D Systems™ Supplier No. 2947CL050

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to bind Jurkat human acute T cell leukemia cells in a flow cytometry assay. A 2-8 fold increase in fluorescence intensity can be observed when Jurkat cells are stained with 10 μg/tube rhLSECtin/his and mouse anti-polyHistidine-PE as compared to cells stained with Mouse Anti-polyHistidine PE-conjugated Monoclonal Antibody (Catalog # IC050P ). Optimal dilutions should be determined by each laboratory for each application.

Specifications

Accession Number NP_940894
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris-Citrate and NaCl.
Gene ID (Entrez) 339390
Molecular Weight (g/mol) M.W. (Observed): 37 kDa, reducing conditions; M.W. (theoretical): 27.8 kDa
Name LSECtin/CLEC4G
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human LSECtin/CLEC4G protein Ser54-Cys293, with an N-terminal 9-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol CLEC4G
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Product Line R&D Systems
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein LSECtin/CLEC4G
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.