Learn More
R&D Systems™ Human Methylated SUMO1 Recombinant Protein
Description
Specifications
Specifications
| Formulation | No Formulation Info in Bulk Inventory |
| Gene ID (Entrez) | 7341 |
| Name | Methylated SUMO1 |
| Quantity | 500μg |
| Source | E. coli-derived human SUMO1 protein |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.