Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Methylcellulose Complete Media Without Epo
SDP

Catalog No. HSC004 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HSC004 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. HSC004 Supplier R&D Systems™ Supplier No. HSC004
Add to Cart
Add to Cart
This item is not returnable. View return policy

For the differentiation and enumeration of granulocyte-macrophage progenitor cells and for detection of Epo-independent erythropoiesis, optimized with cytokines

Successful growth and enumeration of cell colonies is dependent on factors such as accurate cell counts, the presence of growth factors and/or cytokines, adequate humidity, and the use of high quality media.

  • Can be used to identify Epo-independent stimulators of erythropoiesis
  • Optical clarity facilitates colony identification
  • High lot-to-lot consistency decreases variation
  • Supports reproducible in vitro growth of myeloid progenitor cells
  • Does not require addition of growth factors or cytokines
  • Increased cloning efficiency and improved colony growth compared to agar

Specifications

Product Type Methylcellulose Complete Media
Quantity 100 mL
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.