Learn More
R&D Systems™ Human Methylcellulose Serum-Free Enriched Media Without Epo
SDP
Catalog No. HSC010SF Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
HSC010SF 100 mL
1 options
This item is not returnable. View return policy
Catalog No. HSC010SF Supplier R&D Systems™ Supplier No. HSC010SF
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Complete, defined media for the differentiation and enumeration of myeloid progenitor cells and for the detection of Epo-independent erythropoiesis
Specifications
Product Type Methylcellulose Serum-Free Enriched Media
Quantity 100 mL
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.