Description
Includes
ITS Supplement, N-2 MAX Supplement, Bovine Fibronectin Stock, Human FGF basic, Mouse Shh-N, Mouse FGF-8b
The loss of dopaminergic neurons in the substantia nigra is a characteristic of Parkinson's disease (PD), a neurodegenerative disorder that affects motor function.
Pre-optimized kit reduces experimental variation
Defined conditions decreases variation
Includes a panel of premium quality growth factors
Specifications
Specifications
| Quantity | 1 Kit |
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |