Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human/Mouse/Rat Activin A Protein
GSA_VASDP

Catalog No. 338AC010 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
10 μg
50 μg
Formulation:
Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
338AC010 10 μg Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein.
338AC01M 1 mg Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein.
338AC01MCF 1 mg Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
338AC050CF 50 μg Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
338AC050 50 μg Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein.
338AC010CF 10 μg Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 338AC010 Supplier R&D Systems™ Supplier No. 338AC010

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CHO derived

Measured by its ability to induce hemoglobin expression in K562 human chronic myelogenous leukemia cells. Schwall, R.H. et al. (1991) Method Enzymol. 198:340. The ED50 for this effect is 0.2-1.2 ng/mL.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Accession Number P08476
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein.
Gene ID (Entrez) 3624
Molecular Weight (g/mol) M.W. (Observed): 14 kDa, reducing conditions 24 kDa, non-reducing conditions; M.W. (theoretical): 13 kDa (monomer)
Name Activin A
Quantity 10 μg
Source Chinese Hamster Ovary cell line, CHO-derived Activin A protein Gly311-Ser426
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol INHBA
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100-500 μg/mL in sterile 4 mM HCl.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE with silver staining.
Protein Activin A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.