Learn More
R&D Systems™ Human/Mouse/Rat Neural Lineage Functional Identification Kit
SDP
Catalog No. SC028 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
SC028 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. SC028 Supplier R&D Systems™ Supplier No. SC028
This item is not returnable. View return policy
Includes
Neural Maintenance Supplement (10X), Neural Differentiation Supplement, Bovine Fibronectin (100X), NPC Marker: Goat Anti-Rat Nestin Ag Affinity-purified Polyclonal Ab, Astrocyte Marker: Sheep Anti-Human GFAP Ag Affinity-purified Polyclonal Ab, Neuron Mark
To verify the multipotency of neural progenitor cells (NPCs) by in vitro functional differentiation
The ability to verify multipotency at an early stage in the experimental design provides valuable insight that may prevent weeks of wasted effort and reagents. Moreover, confirmation of multipotency helps to ensure consistency among studies and reduce unw
Uses a specially formulated supplement to drive reproducible differentiation
Verifies a healthy, multipotent starting NPC population to increase consistency between studies and reduce unwanted experimental variability
Provides results in 7 to 10 days to
Specifications
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 1 Kit